Learn More
Pluronic™ F-68 Polyol, MP Biomedicals™
Catalog No. MP92750016
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
MP92750016 100 g
1 options
Catalog No. MP92750016 Supplier MP Biomedicals Inc Supplier No. 092750016
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Nonionic detergent suitable for cell culture and insect cell culture.
Pluronic F-68 Polyol is a nonionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
Specifications
Quantity 100 g