missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PluriSln 1

Catalog No. 484710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
484710 10 mg
484750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 484710 Supplier Tocris Bioscience™ Supplier No. 4847/10
Only null left

SCD-1 inhibitor

Stearoyl-CoA desaturase 1 (SCD1) inhibitor. Used to selectively eliminate undifferentiated human pluripotent stem cells (hPSCs) from culture; induces ER stress, attenuates protein synthesis and induces apoptosis in hPSCs. Prevents teratoma formation in immunocompromised mice.

Chemical Identifiers

CAS 91396-88-2
Molecular Formula C12H11N3O

Specifications

Quantity 10 mg
Formula Weight 213.24
Purity >99%
Molecular Weight (g/mol) 213.24
Chemical Name or Material PluriSln 1
Recommended Storage Store at +4°C