accessibility menu, dialog, popup

UserName

ONHHNN

CAS RN 91396-88-2

IUPAC Name: N'-phenylpyridine-4-carbohydrazide
Synonyms: N'-phenylpyridine-4-carbohydrazide
Molecular Weight (g/mol): 213.24
Molecular Formula: C12H11N3O
InChi Key: HUDWXDLBWRHCKO-UHFFFAOYSA-N
SMILES: O=C(NNC1=CC=CC=C1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results