missing translation for 'onlineSavingsMsg'
Learn More

PluriSIn 1 98.0+%, TCI America™

Catalog No. I097520MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I097520MG 20 mg
I0975100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I097520MG Supplier TCI America Supplier No. I097520MG
Only null left

Chemical Identifiers

CAS 91396-88-2
Molecular Formula C12H11N3O
Molecular Weight (g/mol) 213.24
MDL Number MFCD02344904
InChI Key HUDWXDLBWRHCKO-UHFFFAOYSA-N
Synonym plurisln 1, n'-phenylisonicotinohydrazide, plurisln 1 nsc 14613, plurisin 1, plurisin #1 nsc 14613, plurisin #1, 4-pyridinecarboxylicacid, 2-phenylhydrazide, 4-pyridinecarboxylic acid 2-phenylhydrazide, plurisln, plurisln1
PubChem CID 225362
IUPAC Name N'-phenylpyridine-4-carbohydrazide
SMILES O=C(NNC1=CC=CC=C1)C1=CC=NC=C1

Specifications

Melting Point 189°C
Quantity 20 mg
Formula Weight 213.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material PluriSIn 1
TSCA No
Recommended Storage Freezer (-20 °C)