missing translation for 'onlineSavingsMsg'
Learn More

Plumbagin 98.0+%, TCI America™

Catalog No. P11391G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11391G Supplier TCI America Supplier No. P11391G

Chemical Identifiers

CAS 481-42-5
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00001682
InChI Key VCMMXZQDRFWYSE-UHFFFAOYSA-N
Synonym plumbagin, plumbagine, 5-hydroxy-2-methyl-1,4-naphthoquinone, plumbaein, plumbagone, 2-methyljuglone, 5-hydroxy-2-methyl-1,4-naphthalenedione, 2-methyl-5-hydroxy-1,4-naphthoquinone, 5-hydroxy-2-methyl-1,4 naphthoquinone, 1,4-naphthalenedione, 5-hydroxy-2-methyl
PubChem CID 10205
ChEBI CHEBI:8273
IUPAC Name 5-hydroxy-2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=C(C1=O)C=CC=C2O

Specifications

Melting Point 77°C
Quantity 1 g
UN Number 2923
Formula Weight 188.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Plumbagin
RTECSNumber QL8500000
TSCA No