missing translation for 'onlineSavingsMsg'
Learn More

Pleconaril, Thermo Scientific™

Catalog No. AC469221000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469221000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC469221000 Supplier Thermo Scientific Chemicals Supplier No. 469221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 153168-05-9
Molecular Formula C18 H18 F3 N3 O3

Specifications

Quantity 100 mg
Melting Point 55°C to 65°C
Formula Weight 381.35
Physical Form Powder
Color White to Beige
Purity ≥97.5%
Chemical Name or Material Pleconaril
Hazard Category Skull and crossbones
Recommended Storage Normal conditions