accessibility menu, dialog, popup

UserName

H3COCH3H3CNONFFFON

CAS RN 153168-05-9

IUPAC Name: 3-{3,5-dimethyl-4-[3-(3-methyl-1,2-oxazol-5-yl)propoxy]phenyl}-5-(trifluoromethyl)-1,2,4-oxadiazole
Synonyms: pleconaril picovir
Molecular Weight (g/mol): 381.36
Molecular Formula: C18H18F3N3O3
InChi Key: KQOXLKOJHVFTRN-UHFFFAOYSA-N
SMILES: CC1=NOC(CCCOC2=C(C)C=C(C=C2C)C2=NOC(=N2)C(F)(F)F)=C1
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)