Learn More
Invitrogen™ Platinum™ SuperFi II DNA Polymerase
Description
Invitrogen Platinum SuperFi II DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with an innovative buffer, enabling universal primer annealing for the highest success in PCR. Due to the unique composition of the SuperFi II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules.
Features of Platinum SuperFi II DNA Polymerase include:
- Exceptional >300X Taq fidelity
- Universal primer annealing at 60°C
- Superior specificity, sensitivity, and yields
- Robust amplification of difficult-to-amplify targets
Platinum SuperFi II DNA Polymerase is an engineered enzyme with high processivity and increased resistance to PCR inhibitors. It also enables fast-cycling protocols and amplification of long targets (up to 20 kb). Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation. This technology also enables reaction setup at room temperature and provides increased sensitivity and yield.
Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize the annealing temperature for each primer pair. With Platinum SuperFi II DNA Polymerase, different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.
Applications:
- Ultra High-fidelity PCR
- Template generation for sequencing
- CRISPR gene-editing verification and characterization
- Amplification from low-input or suboptimal-purity DNA
- Cloning and construct generation
- Microbial genomics and microbiome research
- Linear template generation for in vitro transcription and recombinant protein expression
- Long-range PCR
- Challenging GC-rich and AT-rich target amplification
- Rare variant and genetic diversity studies
- Multiplex target amplification
- Synthetic biology and DNA assembly
- Site-directed mutagenesis
Specifications
Specifications
| Content And Storage | • Platinum SuperFi II DNA Polymerase (500 μL) • 5X SuperFi II Buffer (6 x 1.25 mL) Store at –20°C. |
| GC-Rich PCR Performance | High |
| Polymerase | Platinum SuperFi II DNA Polymerase |
| Reaction Speed | Fast |
| Quantity | 500 reactions |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Hot-start PCR, High-fidelity PCR |
| Fidelity (vs. Taq) | >300X |
| Hot Start | Built-In Hot Start |
| No. of Reactions | 500 Reactions |
| Show More |
Frequently Asked Questions (FAQs)
Platinum SuperFi II DNA Polymerase enables amplification of long targets (up to 20 kb).
You may want to consider using our Platinum SuperFi II DNA Polymerase (Cat. No. 12361010) and try optimizing your PCR conditions if needed. The Platinum SuperFi II DNA Polymerase demonstrates a superior detection sensitivity with as little as 0.4 ng of genomic DNA. It is also a good idea to include a positive control reaction in parallel to your test PCR to ensure there are no issues with your reagents during handling or use over time. Please note that control templates may be ordered through our GeneArt Gene Synthesis (de novo sequence with 100% guarantee delivered in a vector) or Strings (up to 3 Kb, blunt-ended dsDNA pools, cloned into ZeroBlunt TOPO vector, convenient for multiple controls) services.
Good lab practices are important for long fragment amplification. These include using high-quality templates (pure, fresh, and intact) and fresh primer solutions. Reducing primer concentration to 0.2 µM may also improve the results.
Yes, Platinum SuperFi II DNA Polymerase works at both universal 60 degrees C annealing temperature and at annealing temperatures calculated with a Tm calculator.
Platinum SuperFi II DNA Polymerase produces blunt-ended PCR products that can be cloned directly into blunt-ended cloning vectors. TA cloning is also possible if 3' dA-overhangs are added after PCR. We recommend purifying the PCR products of Platinum SuperFi DNA Polymerase before adding the overhangs. The procedure for adding 3' dA-overhangs (TA cloning) includes the following steps:
- Purify the PCR product (e.g., with a PCR purification kit or phenol extraction/DNA precipitation). Before adding the overhangs, PCR product must be purified, as the strong proofreading activity of any remaining Platinum SuperFi DNA Polymerase will degrade the added 3' dA-overhangs.
- Perform 3' dA addition with a Taq DNA polymerase.
Reaction components:
Purified PCR product
0.2 mM dATP
1x Taq Buffer
1 U Taq DNA Polymerase
Incubate the reaction for 20 min at 72 degrees C.
- Proceed to TA cloning. For optimal ligation efficiency, we recommend using fresh PCR products, since 3' dA-overhangs can be lost during storage