missing translation for 'onlineSavingsMsg'
Learn More

Platinum standard solution, for AAS, 1 mg/ml Pt in 10-20% HCl

Catalog No. AC196191000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196191000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196191000 Supplier Thermo Fisher Scientific Formerly Acros Organics Supplier No. 196191000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy

Specifications

Chemical Name or Material Platinum standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Pt in 10% HCl
CAS 7440-06-4, 7647-01-0, 7732-18-5
Physical Form Solution
Molecular Formula Pt
Concentration 995 to 1005 μg/mL of Pt in 10% HCl (20°C)
Linear Formula Pt
MDL Number MFCD00011179
Solubility Soluble in water
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
Quantity 100 mL
PubChem CID 23939
ChEBI CHEBI:33400
Grade AAS
Show More Show Less

RUO – Research Use Only