missing translation for 'onlineSavingsMsg'
Learn More

Platinum Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLPT32X

 View more versions of this product

Catalog No. PLPT3-2X


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pt1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Platinum Standard
7440-06-4 , 7647-01-0 , 7440-06-4
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,10.0,89.9
MFCD00011179
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
BASFCYQUMIYNBI-UHFFFAOYSA-N
10% HCl
195.08
23939
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,10.0,89.9
Pt
AA, ICP
Certified Reference Material
[Pt]
platinum
500 mL
CHEBI:33400
Matrix Concentration Cat. No.
10% HCl 1000μg/mL PLPT3-2Y, -2X

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
CHEBI:33400
[Pt]
Pt
MFCD00011179
23939
platinum
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only