missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum(II) chloride, 98%
Description
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond, and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond, and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).
Solubility
Soluble in ammonia and hydrochloric acid. Insoluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond, and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).
Solubility
Soluble in ammonia and hydrochloric acid. Insoluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 581°C (decomposition) |
| Quantity | 5 g |
| Density | 5.87 |
| Linear Formula | PtCl2 |
| UN Number | UN3260 |
| Merck Index | 14,7528 |
| Solubility Information | Soluble in ammonia and hydrochloric acid. Insoluble in water,alcohol and ether. |
| Formula Weight | 266 |
| Percent Purity | 98% |
| Chemical Name or Material | Platinum(II) chloride |
Safety and Handling
| Hazard Category | H314-H317-H334 |
| Hazard Statement | GHS H Statement H334-H314-H318-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 233-034-1 |
| RTECSNumber | TP2275000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only