missing translation for 'onlineSavingsMsg'
Learn More

Platinum ICP Standard, 10,000 ppm Pt in 20% HCl, Ricca Chemical

Catalog No. PPT10KH100
Change view
Click to view available options
Emballage:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Emballage
PPT10KH100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PPT10KH100 Supplier Ricca Chemical Company Supplier No. PPT10KH100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-06-4 , 7647-01-0 , 7732-18-5
Formule moléculaire Pt
Poids moléculaire (g/mol) 195.08
Clé InChI BASFCYQUMIYNBI-UHFFFAOYSA-N
PubChem CID 23939
ChEBI CHEBI:33400
Nom de l’IUPAC platinum
SOURIRES [Pt]

Specifications

Nom ou substance chimique Platinum ICP Standard
Emballage Natural LDPE Bottle
Notes sur la concentration ou la composition 10,000ppm Pt
CAS % minimum 68.0, 24.11, 0.68
CAS % maximal 71.0, 25.09, 0.71
Équivalent 1 mL = 10 mg Pt