missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PLATEMASTERTM Height Stop

Catalog No. F1077605
Change view
Click to view available options
For Use With (Equipment):
PLATEMASTER™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
F1077605 PLATEMASTER™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1077605 Supplier Gilson™ Supplier No. F1077605

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easily adjust pipetting height with this pipetting head height adjuster.

The Height Stop is an accessory for the PLATEMASTERTM allowing for easy altering of pipetting height.

  • Ensures appropriate tip depth within shallow- or deep-well microplates

Specifications

Product Type Pipetter Accessory
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Color Black
Material Polypropylene
For Use With (Application) Liquid Handling
For Use With (Equipment) PLATEMASTER™
Quantity 1
Warranty 1 Year
Width (English) 2.76 in
Height (English) .51 in
Height (Metric) 13 mm
Weight (English) .07 lb
Weight (Metric) .03 kg
No. per Pack 1
Show More Show Less