missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Plastic Squeegee

Catalog No. 19263618
Change view
Click to view available options
Color:
Dark Blue
Dark Green
White
Material:
Rubber Blade, Polypropylene Frame
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
19263618 Dark Blue Rubber Blade, Polypropylene Frame
19263619 Dark Green Rubber Blade, Polypropylene Frame
19263617 White Rubber Blade, Polypropylene Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19263618 Supplier Micronova™ Supplier No. SQDB24R

These durably constructed squeegees are made with a polypropylene frame and a choice of either solid rubber or double foam blades for use in wash and production areas. Designed with an ACME threaded handle connection for use with most threaded plastic handles.

The Micronova Plastic Squeegees easily clean smooth or uneven surfaces and minimize contamination areas by reducing dirt and bacteria growth. Designed for use with STAH or STAEH series plastic handles. Available in a variety of colors.

  • Durably constructed polypropylene squeegees
  • Choice of solid rubber or double foam blades
  • Excellent for use on smooth surfaces and uneven floor surfaces
  • Prevents hazardous sparks when cleaning-up flammable fluids
  • Available in 18 and 24 inch (45 and 60 cm) widths
  • Use with STAH or STAEH handles for a secure fit
  • Will not rust

Specifications

Product Type Squeegee, ACME Threaded End
Color Dark Blue
Content And Storage Store in cool dry area when not in use.
Dimensions (L x W) 61 x 5.1 cm (24 x 2 in.)
Disposable Reusable
For Use With (Application) Squeegee for Floors, Walls and Windows
For Use With (Equipment) STA Series Plastic Handles
Material Rubber Blade, Polypropylene Frame
Width (English) 2 in.
Width (Metric) 5.1 cm