missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Plastic Sign Holder: Plastic Holder for 17W x 11H in. Evacuation Map

Catalog No. 19112812
Change view
Click to view available options
Dimensions (L x W):
17.5 x 13.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19112812 17.5 x 13.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19112812 Supplier Brady™ Supplier No. 102849

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve workplace safety with proper signage and markings. This Brady™ plastic sign holder is a durable and effective tool to raise awareness to workplace hazards.

Specifications

Material Plastic
Dimensions (L x W) 17.5 x 13.5 in.
Displayed Text KEEP CYLINDERS CHAINED/MANTENER LOS CILINDROS ENCADENADOS
Color White on Red