Learn More
Innovative Med Tech Plastic Blood Gas Capillary Tubes: Closing Caps
Catalog No. 22757155
Change view
Click to view available options
Diameter (Metric) Outer:
2.0 to 2.4 mm
1.75 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
22757155 2.0 to 2.4 mm
22757150 1.75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22757155 Supplier Innovative Med Tech Supplier No. 110181
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Closing caps for Innovative Med Tech Plastic Blood Gas Capillary Tubes
  • Made from 100% plastic
  • Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Compliance
Meets FDA, OSHA, and CDC mandates
Specifications
Product Type Closing Cap
Material Plastic
For Use With (Equipment) Plastic Blood Gas Capillaries
For Use With (Application) Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Diameter (Metric) Outer 2.0 to 2.4 mm
Certifications/Compliance FDA; OSHA; CDC