missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Plastic Beakers
Plastic Beakers feature excellent clarity and very good chemical resistance.
Supplier: United Scientific™ BP2000PK12
Description
- Molded in polypropylene
- Autoclavable
- Feature raised graduations on the outer surface
- Griffin Style Beakers nest without sticking together
- New Style beakers feature tapered spouts for easier pouring of solutions
Identifiants chimiques
| Polypropylene | |
| Autoclavable | |
| Griffin |
| Beaker | |
| Yes |
Spécifications
| 67.6 oz. | |
| Polypropylene | |
| 12 Pk. | |
| Autoclavable | |
| Yes | |
| Griffin |
| 2000 mL | |
| Beaker for liquid measurement | |
| Beaker | |
| Reusable | |
| Molded Scale |