Learn More
Norgen Biotek Corp Plasma/Serum RNA Purification Midi Kit
Numéro de catalogue. NC1557182
Encompass
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating and exosomal RNA using a two column method. This kit can purify RNA from fresh or frozen serum or plasma samples prepared from blood collected on either EDTA or Citrate, from volumes ranging from 250 L to 1.5 mL. Plasma samples prepared from blood collected on heparin should not be used.The first column will handle the large volume input of bodily fluids that is followed by a concentration on a mini column for a final elution of 50 L to 100 L. All components for the purification and concentration are provided in one convenient and fast kit for the easy processing of large input volumes of bodily fluids.The purified plasma/serum RNA is fully compatible with all downstream applications including NGS and more
Numéro de catalogue. NC1557182 Fournisseur Norgen Biotek Corp Code fournisseur 56100
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating and exosomal RNA using a two column method. This kit can purify RNA from fresh or frozen serum or plasma samples prepared from blood collected on either EDTA or Citrate, from volumes ranging from 250 L to 1.5 mL. Plasma samples prepared from blood collected on heparin should not be used.The first column will handle the large volume input of bodily fluids that is followed by a concentration on a mini column for a final elution of 50 L to 100 L. All components for the purification and concentration are provided in one convenient and fast kit for the easy processing of large input volumes of bodily fluids.The purified plasma/serum RNA is fully compatible with all downstream applications including NGS and more