Learn More
Plasma Emission Standards Kit 1 For ICP, SPEX CertiPrep™
Assurance™ Grade Standards Kit for AA & ICP, 1,000 μg/mL. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Supplier:  SPEX CertiPrep Group LLC ICPKIT1
Numéro de catalogue. ICP-KIT-1
Il en reste null
Découvrez plus d'offres spéciales
Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
  • Assurance™ Grade Standard Kit for AA & ICP
  • 1,000 μg/mL Concentration
  • 125 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Spécifications
Spécifications
Plasma Emission Standards Kit 1
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
38 x 125 mL
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
Each component in a separate 100mL bottle at 1000μg/mL.Component in 20% HCl (in PTFE bottle):TinComponents in 10% HNO3:BismuthMercuryComponents in 2% HNO3:AluminumArsenicBariumBerylliumCadmiumCalciumChromiumCobaltCopperIronLeadLithiumMagnesiumManganeseNickelPotassiumScandiumSeleniumSilverSodiumStrontiumThalliumVanadiumYttriumZincZirconiumComponents in H2O:AntimonyBoronMolybdenumNiobiumPhosphorusSiliconSulfurTitaniumTungsten
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Product intended for laboratory use only