missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2G4E Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA559428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA559428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA559428 Fournisseur Invitrogen™ Code fournisseur PA559428
Il en reste null

Rabbit Polyclonal Antibody

Immunogen sequence: LCLLDTAFFV NSSYPPLLRP ERKADLIIHL NYCAGSQTKP LKQTCEYCTV QNIPFPKYEL PDENENLKEC YLMENPQEPD APIVTFFPLI N Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.

This gene encodes a member of the cytosolic phospholipase A2 group IV family. Members of this family are involved in regulation of membrane tubule-mediated transport. The enzyme encoded by this member of the family plays a role in trafficking through the clathrin-independent endocytic pathway. The enzyme regulates the recycling process via formation of tubules that transport internalized clathrin-independent cargo proteins back to the cell surface.

Spécifications

Antigen PLA2G4E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pla2g4e
Gene Accession No. Q3MJ16
Gene Alias 2310026J01Rik; C230096D22; cPLA2-epsilon; cytosolic phospholipase A2 epsilon; EC 3.1.1.4; Phospholipase A2 group IVE; phospholipase A2, group 4E; phospholipase A2, group IVE; Pla2epsilon; Pla2g4e; RGD1310595
Gene Symbols Pla2g4e
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLA2G4E. Recombinant protein control fragment (Product #RP-98179).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123745
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats