missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PJ 34 hydrochloride

Catalog No. 325550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
325550 50 mg
325510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 325550 Supplier R&D Systems™ Supplier No. 3255/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Potent PARP inhibitor

Potent inhibitor of poly(ADP-ribose) polymerase (PARP) (EC50 = 20 nM). ∽1000-fold more potent than 3-Aminobenzamide (Cat. No. 0788). Protects primary neuronal cells from oxygen-glucose deprivation in vitro and reduces infarct size following focal cerebral ischemia in vivo. Displays protective effects against cisplatin -induced kidney injury. Also displays activity at Pim-1 and Pim-2 kinases at higher concentrations (IC50 values are 3.7 and 16 μM respectively).

Chemical Identifiers

CAS 344458-15-7
Molecular Formula C17H18ClN3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 331.8
Purity >98%
Molecular Weight (g/mol) 331.8
Chemical Name or Material PJ 34 hydrochloride
Recommended Storage Desiccate at Room Temperature