CAS RN 344458-15-7
IUPAC Name:
2-(dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride
Synonyms:
2-(dimethylamino)-N-(6-oxo-5H-phenanthridin-2-yl)acetamide hydrochloride
Molecular Weight (g/mol):
331.8
Molecular Formula:
C17H18ClN3O2
InChi Key:
RURAZZMDMNRXMI-UHFFFAOYSA-N
SMILES:
Cl.CN(C)CC(=O)NC1=CC=C2NC(=O)C3=CC=CC=C3C2=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals PARP Inhibitor VIII, PJ34
CAS: 344458-15-7 Molecular Formula: C17H17N3O2·HCl Synonym: 2-Dimethylamino-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride
2
Tocris Bioscience™ PJ 34 hydrochloride
CAS: 344458-15-7 Molecular Formula: C17H18ClN3O2