missing translation for 'onlineSavingsMsg'
Learn More

Pizotifen 98.0+%, TCI America™

Catalog No. P2344200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2344200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2344200MG Supplier TCI America Supplier No. P2344200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15574-96-6
Molecular Formula C19H21NS
Molecular Weight (g/mol) 295.444
MDL Number MFCD00864192
InChI Key FIADGNVRKBPQEU-UHFFFAOYSA-N
Synonym 4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran
PubChem CID 27400
ChEBI CHEBI:50212
IUPAC Name 4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methylpiperidine
SMILES CN1CCC(=C2C3=C(CCC4=CC=CC=C42)SC=C3)CC1

Specifications

Melting Point 149°C
Color White
Quantity 200 mg
Formula Weight 295.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pizotifen
RTECSNumber TM7165000
TSCA No