missing translation for 'onlineSavingsMsg'
Learn More

Pivalamide 98.0+%, TCI America™

Catalog No. P081925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P081925G 25 g
P0819100G 100 g
P0819500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P081925G Supplier TCI America Supplier No. P081925G

Chemical Identifiers

CAS 754-10-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00008011
InChI Key XIPFMBOWZXULIA-UHFFFAOYSA-N
Synonym pivalamide, trimethylacetamide, 2,2,2-trimethylacetamide, propanamide, 2,2-dimethyl, 2,2-dimethylpropionamide, 2,2-dimethyl-propionamide, unii-fes86mr7zi, fes86mr7zi, propanamide,2,2-dimethyl, trimethylacetarnide
PubChem CID 12957
IUPAC Name 2,2-dimethylpropanamide
SMILES CC(C)(C)C(N)=O

Specifications

Melting Point 154°C
Color White-Yellow
Boiling Point 212°C
Quantity 25 g
Formula Weight 101.15
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pivalamide