Learn More
Fisherbrand™ Pipette-Specific Tips
Tips for specific applications
$96.00 - $210.61
| Catalog Number | Format | Volume (Metric) | Color | Sterility | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Format | Volume (Metric) | Color | Sterility | Price | Quantity & Availability | |||
02707121
![]() |
Rack | 1 to 200 μL | Yellow | Non-sterile |
Pack of 960 Each for $152.29
Case of 5 Pack for $761.45
|
|
|||
02707123
![]() |
Rack | 100 to 1000 μL | Blue | Non-sterile |
Pack of 960 Each for $190.96
Case of 5 Pack for $954.80
|
|
|||
02707124
![]() |
Rack | 100 to 1000 μL | Blue | Sterile |
Pack of 1000 Each for $210.61
Case of 5 Pack for $1,053.05
|
|
|||
21375D
![]() |
Bulk | 1 to 200 μL | Yellow | Non-sterile |
Pack of 1000 Each for $96.00
Case of 10 Pack for $960.00
|
|
|||
21374D
![]() |
Rack, SR 5 x 200 | 1 to 200 μL | Yellow | Non-sterile |
Pack of 1000 Each for $166.79
Case of 10 Pack for $1,667.90
|
|
|||
|
21375E
|
Bulk | 100 to 1000 μL | Blue | Non-sterile |
Pack of 1000 Each for $105.57
Case of 10 Pack for $1,055.70
|
|
|||
Description
These tips are specifically designed to fit certain pipettes and are non-sterile for laboratory use.
- Tips for specific applications
- Designed to fit distinctive pipettes
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free