Learn More
Fisherbrand™ Pipette-Specific Tips
Tips for specific applications
Description
These tips are specifically designed to fit certain pipettes and are non-sterile for laboratory use.
- Tips for specific applications
- Designed to fit distinctive pipettes
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
| 100 to 1000 μL | |
| Blue | |
| 7.1cm | |
| Specialty |
| Sterile | |
| 2.8 in. | |
| Rack | |
| Eppendorf |