Learn More
Fisherbrand™ Pipette-Specific Tips
GREENER_CHOICE
Catalog No. 02707121
Change view
Click to view available options
Format:
Bulk
Rack
Rack, SR 5 x 200
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Yellow
Sterility:
Non-sterile
Sterile
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Volume (Metric) Color Sterility
02707121 Rack 1 to 200 μL Yellow Non-sterile
02707123 Rack 100 to 1000 μL Blue Non-sterile
02707124 Rack 100 to 1000 μL Blue Sterile
21375D Bulk 1 to 200 μL Yellow Non-sterile
21374D Rack, SR 5 x 200 1 to 200 μL Yellow Non-sterile
21375E Bulk 100 to 1000 μL Blue Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02707121
Tips for specific applications
These tips are specifically designed to fit certain pipettes and are non-sterile for laboratory use.
  • Tips for specific applications
  • Designed to fit distinctive pipettes
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Yellow
Length (English) 1.88 in.
Length (Metric) 4.78cm
Format Rack
For Use With (Equipment) Corning Lambda, Eppendorf Reference, Eppendorf Research, Gilson Pipetman, Labpette Digital, Labpette Digital, Mettler Toledo VoluMate, Nichiryo, Oxford Benchmate, Gilson Pipetman, Socorex, Thermo Scientific Finnpipette, Rainin Classic