missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAXTM Tip Reload Block

Catalog No. 32000175
Change view
Click to view available options
For Use With (Equipment):
PIPETMAX
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
32000175 PIPETMAX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32000175 Supplier Gilson™ Supplier No. 32000175

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tip reloading block for use with its automated pipettor.

Reload your pipettor tips with ease with the PIPETMAXTM Tip Reload Block.

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Color White
Material Polypropylene
For Use With (Application) Liquid Handling
For Use With (Equipment) PIPETMAX
Warranty 1 Year