missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Pipet Stands and Accessories
Description
Pipette Stands
- Stores pipettes in an upright position while not in use to avoid contamination from the desktop
- Linear Stand is designed for all mechanical devices and stores them neatly in a line for instant access
- Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
- mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
- Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
- Includes 5 color-coded replacement caps
- Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
- Can also be used as a tube opener
Spécifications
Spécifications
| Color | White/Black |
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Pipette Holder |
| For Use With (Application) | Storing pipets when not in use |
| For Use With (Equipment) | mLINE/Proline Plus Mechanical Pipette |
| Quantity | 1 Each |
| Dimensions (L x W x H) | 29mmW x 250mmD x 60mmH |