missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219483880
Specifications
| PIPES | |
| White | |
| 100 g | |
| C8H18N2O6S2 | |
| Piperazine-N, N'-bis[2-ethanesulfonic acid] | |
| C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O | |
| 302.36 | |
| CHEBI:44933 | |
| Molecular Biology Reagent |
| 5625-37-6 | |
| Powder | |
| ≥99% | |
| MFCD00006159 | |
| IHPYMWDTONKSCO-UHFFFAOYSA-N | |
| 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid | |
| 79723 | |
| 302.4 |
Chemical Identifiers
| 5625-37-6 | |
| 302.36 | |
| IHPYMWDTONKSCO-UHFFFAOYSA-N | |
| 79723 | |
| 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
| C8H18N2O6S2 | |
| MFCD00006159 | |
| Piperazine-N, N'-bis[2-ethanesulfonic acid] | |
| CHEBI:44933 | |
| C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
Safety and Handling
EINECSNumber : 227-057-6