missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
$57.69 - $198.90
Chemical Identifiers
| CAS | 5625-37-6 |
|---|---|
| Molecular Formula | C8H18N2O6S2 |
| Molecular Weight (g/mol) | 302.36 |
| MDL Number | MFCD00006159 |
| InChI Key | IHPYMWDTONKSCO-UHFFFAOYSA-N |
| Synonym | Piperazine-N, N'-bis[2-ethanesulfonic acid] |
| PubChem CID | 79723 |
| ChEBI | CHEBI:44933 |
| IUPAC Name | 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
| SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19483825
|
MP Biomedicals Inc
0219483825 |
25 g |
Each for $57.69
|
|
|||||
|
ICN19483880
|
MP Biomedicals Inc
0219483880 |
100 g |
Each for $198.90
|
|
|||||
Chemical Identifiers
| 5625-37-6 | |
| 302.36 | |
| IHPYMWDTONKSCO-UHFFFAOYSA-N | |
| 79723 | |
| 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
| C8H18N2O6S2 | |
| MFCD00006159 | |
| Piperazine-N, N'-bis[2-ethanesulfonic acid] | |
| CHEBI:44933 | |
| C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
Specifications
| 5625-37-6 | |
| Powder | |
| ≥99% | |
| MFCD00006159 | |
| IHPYMWDTONKSCO-UHFFFAOYSA-N | |
| 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid | |
| 79723 | |
| 302.4 | |
| PIPES |
| White | |
| 25 g | |
| C8H18N2O6S2 | |
| Piperazine-N, N'-bis[2-ethanesulfonic acid] | |
| C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O | |
| 302.36 | |
| CHEBI:44933 | |
| Molecular Biology Reagent |
Safety and Handling
EINECSNumber : 227-057-6