missing translation for 'onlineSavingsMsg'
Learn More

PIPES, 98%

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

Supplier:  Thermo Scientific Chemicals A1609022

 View more versions of this product

Catalog No. AAA1609022


Only null left

Description

Description

PIPES is commonly used as a buffering agent in biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES is commonly used as a buffering agent in biochemistry.

Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

PIPES
5625-37-6
C8H18N2O6S2
817713
pipes, 1,4-piperazinediethanesulfonic acid, piperazine-n,n'-bis 2-ethanesulfonic acid, 2,2'-piperazine-1,4-diyl diethanesulfonic acid, 1,4-piperazinebis ethanesulfonic acid, unii-g502h79v6l, 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonic acid, piperazine-1,4-bis 2-ethanesulfonic acid, 1,4-piperazine-diethanesulfonic acid, 2,2'-piperazine-1,4-diyl bis ethanesulphonic acid
IHPYMWDTONKSCO-UHFFFAOYSA-N
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
79723
302.37
>300°C
100 g
MFCD00006159
147479
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
302.36
CHEBI:44933
98%

Chemical Identifiers

5625-37-6
302.36
IHPYMWDTONKSCO-UHFFFAOYSA-N
79723
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C8H18N2O6S2
MFCD00006159
pipes, 1,4-piperazinediethanesulfonic acid, piperazine-n,n'-bis 2-ethanesulfonic acid, 2,2'-piperazine-1,4-diyl diethanesulfonic acid, 1,4-piperazinebis ethanesulfonic acid, unii-g502h79v6l, 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonic acid, piperazine-1,4-bis 2-ethanesulfonic acid, 1,4-piperazine-diethanesulfonic acid, 2,2'-piperazine-1,4-diyl bis ethanesulphonic acid
CHEBI:44933
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-057-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only