missing translation for 'onlineSavingsMsg'
Learn More

PIPES, 98%

Catalog No. p-7023043
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1609014 25 g
AAA1609022 100 g
AAA1609036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1609014 Supplier Thermo Scientific Chemicals Supplier No. A1609014

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

PIPES is commonly used as a buffering agent in biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES is commonly used as a buffering agent in biochemistry.

Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym pipes, 1,4-piperazinediethanesulfonic acid, piperazine-n,n'-bis 2-ethanesulfonic acid, 2,2'-piperazine-1,4-diyl diethanesulfonic acid, 1,4-piperazinebis ethanesulfonic acid, unii-g502h79v6l, 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonic acid, piperazine-1,4-bis 2-ethanesulfonic acid, 1,4-piperazine-diethanesulfonic acid, 2,2'-piperazine-1,4-diyl bis ethanesulphonic acid
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 817713
Merck Index 147479
Solubility Information Soluble in 1M sodium hydroxide solution. Slightly soluble in water.
Formula Weight 302.37
Percent Purity 98%
Chemical Name or Material PIPES
EINECSNumber 227-057-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only