missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, Anhydrous, 99%, Spectrum™ Chemical

Catalog No. 18612387
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612387 500 g Amber Glass Bottle
18612385 5 g Amber Glass Bottle
18612381 100 g Amber Glass Bottle
18612383 2.5 kg Amber Glass Bottle
18612382 12 kg Amber Glass Bottle
18612384 25 kg Amber Glass Bottle
18612386 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18612387 Supplier Spectrum Chemical Mfg Cor Supplier No. PI123500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

CAS Max % 1
Melting Point 108° to 113°C
Boiling Point 147°C / 296.6°F
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Piperazine, Anhydrous

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.