missing translation for 'onlineSavingsMsg'
Learn More

PIP1/WDR84 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560672
Encompass
Change view
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
501560672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560672 Supplier Bethyl Laboratories Inc Supplier No. A301550A

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Negatively regulates the PAK1 kinase. PAK1 is a member of the PAK kinase family, which have been shown to play a positive role in the regulation of signaling pathways involving MAPK8 and RELA. PAK1 exists as an inactive homodimer, which is activated by binding of small GTPases such as CDC42 to an N-terminal regulatory domain. PAK1IP1 also binds to the N-terminus of PAK1, and inhibits the specific activation of PAK1 by CDC42.

Specifications

Antigène PIP1/WDR84
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugué Unconjugated
Formule TBS with 0.1% BSA and 0.09% sodium azide
Gène PAK1IP1
Numéro d’acquisition du gène Q9NWT1
Alias du gène MAK11, PIP1, RP11-421M1.5, WDR84, bA421M1.5, hPIP1, PAK/PLC-interacting protein 1, PAK1-interacting protein 1, WD repeat-containing protein 84, p21-activated protein kinase-interacting protein 1
Symboles de gène PAK1IP1
Espèces hôtes Rabbit
Immunogène Between 342 and 392
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 55003
Espèces cibles Human
Contenu et stockage 4° C
Type de produit Antibody
Forme Liquid
Isotype IgG
Show More Show Less