missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pioglitazone hydrochloride

Catalog No. 412410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
412410 10 mg
412450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 412410 Supplier Tocris Bioscience™ Supplier No. 4124/10

Selective PPARγ agonist; antidiabetic agent

Pioglitazone hydrochloride is a selective PPARγ agonist (EC50 = 0.69 μM). Thiazolidinedione (TZD) derivative and antidiabetic agent; improves insulin sensitivity. Also enhances proliferation and survival of mESCs and acts synergistically with Y-27632 (Cat. No. 1254) to improve survival of dissociated single hPSCs and increase colony formation.

Chemical Identifiers

CAS 112529-15-4
Molecular Formula C19H21ClN2O3S

Specifications

Quantity 10 mg
Formula Weight 392.9
Purity >99%
Molecular Weight (g/mol) 392.898
Chemical Name or Material Pioglitazone hydrochloride
Recommended Storage Desiccate at Room Temperature