missing translation for 'onlineSavingsMsg'
Learn More

Pilocarpine Nitrate 98.0+%, TCI America™

Catalog No. P03995G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P03995G Supplier TCI America Supplier No. P03995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148-72-1
Molecular Formula C11H17N3O5
Molecular Weight (g/mol) 271.273
MDL Number MFCD00078497
InChI Key PRZXEPJJHQYOGF-GNAZCLTHSA-N
PubChem CID 657349
IUPAC Name (3S,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;nitric acid
SMILES CCC1C(COC1=O)CC2=CN=CN2C.[N+](=O)(O)[O-]

Specifications

Melting Point 171°C
Color White
Quantity 5 g
Formula Weight 271.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pilocarpine Nitrate
RTECSNumber TK1455000
TSCA Yes