missing translation for 'onlineSavingsMsg'
Learn More

Pilocarpine Nitrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019239
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019239 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019239 Supplier Merck Emd Millipore Supplier No. PHR1494500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Pilocarpine Nitrate is a naturally occurring cholinomimetic drug, exhibiting nicotinic and muscarinic properties. It is widely used for controlling intraocular pressure in glaucoma, immediate relief of mydriasis, and to reverse mydriasis caused by cycloplegic agents.

Specifications

CAS 148-72-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C11H16N2O2 · HNO3
MDL Number MFCD00078497
Synonym Pilocarpine nitrate salt
Type Certified Reference Material
Quantity 500 mg
Formula Weight 271.27
Grade Certified Reference Material