missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Pig Dissection Model

Catalog No. S27906
Change view
Click to view available options
Dimensions (L x W x H):
56 x 30 x 7 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S27906 56 x 30 x 7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27906 Supplier Eisco™ Supplier No. ZM16089

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This pig dissection model is a great alternative or supplement to traditional dissection, it is also useful for virtual learning.

  • Model demonstrates the general structure and internal organs of a pig
  • Features include detachable liver, intestines, two piece heart, lungs, and stomach
  • Made of durable polyvinyl chloride (PVC) plastic which is corrosion resistant, lightweight, and has high strength
  • Resists reactions with acids, gasoline, alcohol, and hydrocarbons
  • Vividly hand painted with eco friendly paint
  • An alternative or supplement to traditional dissection in the classroom
  • Identification key card included

Specifications

Dimensions (L x W x H) 56 x 30 x 7 cm
For Use With (Application) Studying Important Exterior/Interior Parts, Organs, and Systems
Includes Identification Key Card
Material PVC
Mounting Base
Weight (Metric) 2.78 kg
Product Type Model