missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated Plates

Catalog No. PI15120
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Plate Blocking:
Blocker™ BSA Buffer
SuperBlock™ Buffer
Quantity:
5 Plates
5 x 5 Plates
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format Plate Blocking Quantity
PI15120 Clear Strip Plate SuperBlock™ Buffer 5 Plates
PI15219 Black Standard Blocker™ BSA Buffer 5 Plates
PI15119 Black Standard SuperBlock™ Buffer 5 Plates
PI15407 Black Standard SuperBlock™ Buffer 5 Plates
PI15121 Clear Strip Plate Blocker™ BSA Buffer 5 Plates
PI15122 Clear Strip Plate SuperBlock™ Buffer 5 x 5 Plates
PI15125 Clear Standard Blocker™ BSA Buffer 5 Plates
PI15124 Clear Standard SuperBlock™ Buffer 5 Plates
PI15126 Clear Standard SuperBlock™ Buffer 5 x 5 Plates
PI15405 Clear Standard SuperBlock™ Buffer 5 Plates
PI15218 White Standard Blocker™ BSA Buffer 5 Plates
PI15118 White Standard SuperBlock™ Buffer 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. PI15120 Supplier Thermo Scientific™ Supplier No. LSG15120

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.

Streptavidin-coated microplates provide the biotin-binding function needed for a variety of ELISA and other assay procedures based on biotinylated capture antibodies, peptide antigens, and other analytes such as oligonucleotides. Pierce Streptavidin Coated Plates are available in clear, white, and black plates for use with colorimetric, chemiluminescent, and fluorescent detection methods, respectively.

Thermo Scientific Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.

Features of Streptavidin Coated Plates include:
• Approximate binding capacity is ∼5 pmol biotin/well
• Easy and gentle immobilization of biotin-containing conjugates
• Low nonspecific binding
• No denaturing of the protein component of a conjugate upon binding
• Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
• Pre-blocked with your choice of Blocker BSA or SuperBlock Blocking Buffer

Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions.

Specifications

Binding Property Standard Capacity, Affinity
Format Strip Plate
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Streptavidin
Plate Blocking SuperBlock™ Buffer
Quantity 5 Plates
Show More Show Less