missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Streptavidin
Description
- Purified streptavidin is supplied lyophilized in phosphate-based buffer for immediate reconstitution with water
- Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii
- Pierce™ streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI
- Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin
- Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation
- Applications: ELISA (assay dependent), western blot (assay dependent)
Spécifications
Spécifications
| Conjugate | Unconjugated |
| Quantity | 1 mg |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. |
| Form | Solid |
| Product Line | Pierce |
| Product Type | Streptavidin |
| Target Molecule | Biotin |
| Label or Dye | Unlabeled |
For Research Use Only. Not for use in diagnostic procedures.