Learn More
Thermo Scientific™ Pierce™ SMCC
No-Weigh™ Format
$327.22 - $438.08
Specifications
Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Molecular Weight (g/mol) 334.32
PEGylated No
Spacer Arm Length 8.3 Å
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI22360
View Documents
Thermo Scientific™
LSG22360
50 mg
Pack for $438.08
Only null left
PIA35394
View Documents
Thermo Scientific™
A35394
10 x 1 mg
Each for $327.22
Only null left
Description
Description
Thermo Scientific™ Pierce SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm (8.3 Ås).
Highlights:
  • Cyclohexane bridge adds stability to the maleimide-reactive group; N-(4-carboxycyclohexylmethyl) maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7.0 at 4°C
  • Ideal for coupling enzymes to antibodies
  • Reactive groups: NHS-ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
Specifications
Specifications
Amine-Sulfhydryl
334.32
8.3 Å
Yes
Chemical Labeling
Maleimide, NHS Ester
Powder
Standard, Single-use
Crosslinker
By Thiols
No
Upon receipt store desiccated at 4°C.
Ambient
Heterobifunctional
Short (<10 Å)
DMF, DMSO
No
Videos
SDS
Documents
Documents
For Research Use Only. Not for use in diagnostic procedures.