missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein L

Catalog No. PI77679
Change view
Click to view available options
Quantity:
1 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77679 50 mg
PI21189 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI77679 Supplier Thermo Scientific™ Supplier No. LSG77679

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect mouse and human antibodies containing kappa light chains in immunoassays using this purified, biotinylated or peroxidase-conjugated, recombinant Protein L.

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein L enable probing and detection of mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Highlights:

  • Contains four immunoglobulin-binding domains per protein
  • Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Specifications

Quantity 50 mg
Product Line Pierce
Form Powder
Protein Subtype Protein L
Target Antibodies
Content And Storage Upon receipt store at 4°C.

Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.