Learn More
Thermo Scientific™ Pierce™ Quantitative Peroxide Assay Kit (Lipid)
Description
Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.
The Thermo Scientific Pierce Quantitative Peroxide Assay Kit (lipid-compatible formulation) detects and measures hydrogen peroxide levels in biological samples using an iron and xylenol orange (XO) reagent.
This lipid-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. The kit's formulation may be used with lipid-containing samples without first extracting the lipids; however, because this kit does not contain sorbitol, the assay is less sensitive. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.
Features of the Quantitative Peroxide Assay Kit:
- Peroxides—detects and measures hydrogen peroxide (H2O2) levels in biological and other liquids samples
- Convenient—no proprietary reagents involved but the ready-to-use kit removes the variability, difficulty and added expense of formulating the reagents yourself
Applications:
- General hydrogen peroxide (H2O2) measurement in protein samples
- Quantitation of peroxides in detergents
- Assessment of the effects of peroxides on cellular activity
- Determination of protein glycation; Measurement of peroxide accumulation in lipids
In this assay, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With this lipid-compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560 nm. Peroxide levels in test samples can be determined by calculation from the known extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.
Related Products
- Pierce™ Quantitative Peroxide Assay Kit (Aqueous) (Cat. No. 23280)
- Pierce™ Peroxide Assay Reagent A (Cat. No. 23281)
- Pierce™ Peroxide Assay Reagent B (Cat. No. 23282)
- Pierce™ Peroxide Assay Reagent C (Cat. No. 23283)
Specifications
Specifications
| Content And Storage | Sufficient For: 100 tube assays or 500 microplate assays with lipid-containing samples • Reagent A, 1 mL • Reagent C, 4 x 25 mL Store at 4°C. |
| Description | Pierce Quantitative Peroxide Assay Kit (Lipid) |
| Assay | Quantitative Peroxide Assay Kit |
| Detection Method | Absorbance |
| Product Line | Pierce |
| Product Type | Quantitative Peroxide Assay |
| Specificity | Not Target-Specific |
| Sufficient For | 100 Tube Assays or 500 Microplate Assays |
| For Use With (Application) | Solution-based Detection, Absorbance |
| For Use With (Equipment) | Spectrophotometer, Microplate Reader |
| Show More |
Frequently Asked Questions (FAQs)
We offer several specialty protein assays including: Protease assay kits (Cat. Nos. 23263 and 23266), Glycoprotein Carbohydrate Estimation Kit (Cat. No. 23260), Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270), Quantitative Peroxide assay kits (Cat. Nos. 23280 and 23285), and Easy-Titer Assay kits for IgG and IgM.
For Research Use Only. Not for use in diagnostic procedures.