missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A

Catalog No. PI21181
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21181 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21181 Supplier Thermo Scientific™ Supplier No. 21181

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect rabbit and human antibodies, especially IgG isotypes, for Western blot, ELISA and other immunoassays with this purified and biotinylated recombinant Protein A.

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein A enable probing and detection of rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

Protein A is a cell wall component produced by several strains of Staphylococcus aureus that consists of a single polypeptide chain and contains little or no carbohydrate. The Protein A molecule contains four high-affinity binding sites capable of interacting with the Fc region from IgG of several species including human and rabbit. Optimal binding occurs at pH 8.2, although binding is also effective at neutral or physiological conditions (pH 7.0 to 7.6).

Highlights:

  • Contains four Fc-binding domains per protein
  • Better than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Poorer than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies

Specifications

Product Type Protein A
Quantity 5 mg
Product Line Pierce
Form Powder
Protein Subtype Protein A
Species Human, Rabbit
Target Antibodies
Content And Storage Upon receipt store at -20°C. Product is shipped with an ice pack.

For Research Use Only. Not for use in diagnostic procedures.