missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets,EDTA-free
Description
The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.
Highlights:- Improved formulation&mdsashtablets dissolve quickly into a clear solution
- Economical – more cost-effective than other commercially available formulations for equivalent volumes
- Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
- Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
- EDTA-free- formulated without EDTA, a metalloproteinase inhibitor
Specifications
Specifications
| Includes | 1 Vial Inhibitor Tablets, EDTA Free |
| Product Type | Protease and Phosphatase Inhibitor |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | protein purification, western blotting |
| Sufficient For | 1 Tablet is Sufficient for 10 mL |
| Quantity | 2 Tablets |