missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets,EDTA-free

Catalog No. PIA32960
Change view
Click to view available options
Quantity:
2 Tablets
Includes:
1 Vial Inhibitor Tablets
1 Vial Inhibitor Tablets, EDTA Free
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Includes
PIA32960 2 Tablets 1 Vial Inhibitor Tablets
PIA32962 2 Tablets 1 Vial Inhibitor Tablets, EDTA Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PIA32960 Supplier Thermo Scientific™ Supplier No. A32960
For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.

The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
* EDTA is present only in Product No. A32959.

Specifications

Includes 1 Vial Inhibitor Tablets
Product Type Protease and Phosphatase Inhibitor
Content And Storage 2°C to 8°C
For Use With (Application) protein purification, western blotting
Sufficient For 1 Tablet is Sufficient for 10 mL
Quantity 2 Tablets