Learn More
Thermo Scientific™ Pepstatin A Protease Inhibitor
Description
Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.
Highlights:
- High-quality reagents in convenient and affordable package sizes
- Purchase individually to prepare customized protease inhibitor cocktails
- Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
Specifications
Specifications
| Description | Pierce Pepstatin A Protease Inhibitor |
| Product Type | Protease Inhibitor |
| Content And Storage | Store between 2°C and 8°C |
| Form | Lyophilized |
| Product Line | Pierce |
| Reagent Type | Protease Inhibitor |
| Inhibitors | Protease Inhibitor |
| Quantity | 25 mg |
Frequently Asked Questions (FAQs)
We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.
Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:
- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).
For Research Use Only. Not for use in diagnostic procedures.