Learn More
Thermo Scientific™ Aprotinin Protease Inhibitor
Description
Thermo Scientific Aprotinin Protease Inhibitor reversably inhibits serine proteases. Thermo Scientific protease inhibitors are small sizes of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
Features of these protease inhibitors include:
- High-quality reagents in convenient and affordable package sizes
- Purchase individually to prepare customized protease inhibitor cocktails
- Supplement Halt Protease Inhibitor Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
The individual reagents offered include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A and PMSF. Although Halt Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.
Note: Aprotinin Protease Inhibitor is readily soluble in water (up to ~10 mg/mL) and compatible with reconstitution in deionized water or a suitable aqueous buffer.
Spécifications
Spécifications
| Description | Pierce Aprotinin Protease Inhibitor |
| Product Type | Protease Inhibitor |
| Content And Storage | Store between 2°C and 8°C |
| Form | Lyophilized |
| Product Line | Pierce |
| Reagent Type | Protease Inhibitor |
| Inhibitors | Protease Inhibitor |
| Quantity | 25 mg |
For Research Use Only. Not for use in diagnostic procedures.