Learn More
Thermo Scientific™ PMSF Protease Inhibitor
Catalog No. PI36978
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
PI36978 5 g
1 options
Catalog No. PI36978 Supplier Thermo Scientific™ Supplier No. LSG36978
Season of Savings
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.
Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.
Highlights:
  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
Specifications
Description Pierce PMSF Protease Inhibitor
Product Type Protease Inhibitor
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Powder
Reagent Type Protease Inhibitor
Inhibitors Protease Inhibitor
Quantity 5 g
How should I reconstitute and store PMSF Protease Inhibitor (Cat. No. 36978)?
We recommend dissolving PMSF Protease Inhibitor (Cat. No. 36978) in isopropanol at 1.74 mg/mL (10 mM) and storing in aliquots at -20 degrees C.
For Research Use Only. Not for use in diagnostic procedures.